Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fas3
Entrez Gene ID
35097
Gene Name
Fasciclin 3
Gene Aliases
CG5803, Dmel\CG5803, Dmel_CG5803, FAS III, FAS3, FASIII, Fas, Fas III, Fas-3, Fas-III, FasIII, FasIIIface, FascIII, fas III, fas-III, fas3, fasIII, fascIII
Chromosome Location
Chr.2L: 18320102 - 18393441 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000636
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103704.3 NP_001097174.1 4 - 5 1730 53
NM_001169534.3 NP_001163005.1 4 - 5 1730 53
NM_001299109.1 NP_001286038.1 4 - 5 1730 53
NM_001299110.1 NP_001286039.1 4 - 5 1730 53
NM_165249.4 NP_724106.1 4 - 5 1730 53
NM_165250.4 NP_724107.1 4 - 5 1730 53
GenBank mRNA AY075516.1 - 4 - 5 1725 53
BT133041.1 - 4 - 5 1709 53
M27813.1 - 4 - 5 1728 53

View Full Product Details