Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mst36Fa
- Entrez Gene ID
-
318948
- Gene Name
- Male-specific transcript 36Fa
- Gene Aliases
-
CG15165, CG31801, Dmel\CG31801, Dmel_CG31801, mst36Fa
- Chromosome Location
-
Chr.2L: 18675183 - 18676998 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086681
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165259.3 |
NP_724121.3 |
2 - 3 |
926 |
81 |
View Full Product Details