Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acn
Entrez Gene ID
35173
Gene Name
Acinus
Gene Aliases
Ba, CG10437, CG10473, Dmel\CG10473, Dmel_CG10473, acn, dAcn, dacn, hkl, l(2)37Ba
Chromosome Location
Chr.2L: 19045355 - 19048817 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263198
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273627.1 NP_001260556.1 3 - 4 2144 72
NM_136091.4 NP_609935.1 4 - 5 2059 72
NM_165273.2 NP_724153.1 3 - 4 2144 72
GenBank mRNA AY069519.1 - 3 - 4 2105 72

View Full Product Details