Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lok
Entrez Gene ID
35288
Gene Name
loki
Gene Aliases
10895, 38B.4, CG10895, CHK-2, CHK2, Chk2, DmCHk2, DmChk2, Dmchk2, Dmel\CG10895, Dmel_CG10895, Dmnk, Dmnk-L, Dmnk-S, Dmnk/DChk2, LOKI/CHK2, Lok, Loki, MNK, Mnk, chk, chk-2, chk2, chk2/lok, mnk
Chromosome Location
Chr.2L: 20061422 - 20064381 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0019686
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259171.2 NP_001246100.1 2 - 3 675 89
NM_057870.4 NP_477218.1 3 - 4 614 89
NM_057871.4 NP_477219.1 3 - 4 740 89
NM_165318.3 NP_724241.1 2 - 3 675 89
GenBank mRNA AY070549.1 - 3 - 4 569 89
BT120173.1 - 2 - 3 642 89
BT120181.1 - 2 - 3 642 89

View Full Product Details