Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spir
Entrez Gene ID
45931
Gene Name
spire
Gene Aliases
38C.34, 38C.37, CG10076, CG18621, Dmel\CG10076, Dmel_CG10076, NP2788, Spir, Spire, l(2)08327, p150-Spir, p150Spir
Chromosome Location
Chr.2L: 20311219 - 20348386 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003475
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169549.1 NP_001163020.1 6 - 7 1695 73
NM_001259172.2 NP_001246101.1 6 - 7 1692 73
NM_001259173.1 NP_001246102.1 6 - 7 1698 73
NM_001259174.1 NP_001246103.1 6 - 7 1692 73
NM_001299171.1 NP_001286100.1 6 - 7 1698 73
NM_080115.3 NP_524854.2 6 - 7 1698 73
NM_165323.3 NP_724254.1 6 - 7 1698 73
NM_165324.2 NP_724255.1 6 - 7 1695 73
GenBank mRNA AJ238876.1 - 6 - 7 1345 73
AY075586.1 - 6 - 7 1684 73
BT122081.1 - 6 - 7 1374 73

View Full Product Details