Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spir
Entrez Gene ID
45931
Gene Name
spire
Gene Aliases
38C.34, 38C.37, CG10076, CG18621, Dmel\CG10076, Dmel_CG10076, NP2788, Spir, Spire, l(2)08327, p150-Spir, p150Spir
Chromosome Location
Chr.2L: 20311219 - 20348386 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003475
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169549.1 NP_001163020.1 9 - 10 2073 66
NM_001259172.2 NP_001246101.1 9 - 10 2070 66
NM_001259173.1 NP_001246102.1 9 - 10 2076 66
NM_001299171.1 NP_001286100.1 9 - 10 2076 66
NM_080115.3 NP_524854.2 9 - 10 2076 66
NM_165323.3 NP_724254.1 9 - 10 2076 66
NM_165324.2 NP_724255.1 9 - 10 2073 66
GenBank mRNA AJ238876.1 - 9 - 10 1723 66
AY075586.1 - 9 - 10 2062 66
BT122081.1 - 9 - 10 1752 66

View Full Product Details