Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spir
Entrez Gene ID
45931
Gene Name
spire
Gene Aliases
38C.34, 38C.37, CG10076, CG18621, Dmel\CG10076, Dmel_CG10076, NP2788, Spir, Spire, l(2)08327, p150-Spir, p150Spir
Chromosome Location
Chr.2L: 20311219 - 20348386 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003475
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259173.1 NP_001246102.1 11 - 12 2350 117
NM_001299171.1 NP_001286100.1 11 - 12 2350 117
NM_165324.2 NP_724255.1 11 - 12 2347 117
GenBank mRNA AY075586.1 - 11 - 12 2336 117
BT122081.1 - 11 - 12 2026 117

View Full Product Details