Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IKKepsilon
Entrez Gene ID
35329
Gene Name
I-kappaB kinase epsilon
Gene Aliases
38D.31, APTX7, CG2615, DIK2, Dik2, DmIKKepsilon, DmIKKepsilon/dIK2, Dmel\CG2615, Dmel_CG2615, Dmik2, IK2, IKK, Ik2, Ik2/DmIKK, dIKK, dIKKepsilon, dik2, ik2, ikkepsilon, l(2)38Ea
Chromosome Location
Chr.2L: 20676045 - 20678979 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086657
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136204.3 NP_610048.2 5 - 6 1645 90
NM_165337.2 NP_724278.1 4 - 5 1555 90
GenBank mRNA BT046140.2 - 4 - 5 1529 90

View Full Product Details