Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Nhe2
- Entrez Gene ID
-
35376
- Gene Name
- Na[+]/H[+] hydrogen exchanger 2
- Gene Aliases
-
CG9255, CG9256, DNhe2, DmNHE2, Dmel\CG9256, Dmel_CG9256, Dnhe2, NHE2, dNhe2, nhe2
- Chromosome Location
-
Chr.2L: 21104568 - 21134255 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040297
- Gene Symbol
- CG46307
- Entrez Gene ID
-
30522396
- Gene Name
- CG46307-PA
- Gene Aliases
-
Dmel\CG46307, Dmel_CG46307, Nhe2-5'utr
- Chromosome Location
-
Chr.2L: 21104568 - 21132909 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0284223
View Full Product Details