Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ac3
- Entrez Gene ID
-
35419
- Gene Name
- Adenylate cyclase 3
- Gene Aliases
-
AC 3, AC3, CG1506, DAC3, DAC39E, Dmel\CG1506, Dmel_CG1506, ac3
- Chromosome Location
-
Chr.2L: 21632587 - 21647964 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0023416
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136272.3 |
NP_610116.2 |
7 - 8 |
1433 |
66 |
View Full Product Details