Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ars2
Entrez Gene ID
35539
Gene Name
Arsenic resistance protein 2
Gene Aliases
CG7843, Dmel\CG7843, Dmel_CG7843, anon-WO0118547.166, ars2
Chromosome Location
Chr.2R: 6080828 - 6085625 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033062
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136359.3 NP_610203.2 2 - 3 220 100
NM_165458.2 NP_724453.1 2 - 3 299 100
NM_165459.2 NP_724454.1 1 - 2 350 100
NM_165460.2 NP_724455.2 2 - 3 220 100
GenBank mRNA BT011368.1 - 2 - 3 229 100

View Full Product Details