Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfk
Entrez Gene ID
36060
Gene Name
Phosphofructokinase
Gene Aliases
BcDNA.GH12192, BcDNA:GH12192, CG4001, Dmel\CG4001, Dmel_CG4001, PFK, PFK1, l(2)06339, pfk
Chromosome Location
Chr.2R: 10109740 - 10117457 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003071
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259305.1 NP_001246234.1 9 - 10 1532 67
NM_001273918.1 NP_001260847.1 8 - 9 1448 67
NM_078952.3 NP_523676.1 9 - 10 1532 67
NM_165746.3 NP_724890.1 5 - 6 1503 67
NM_165747.4 NP_724891.2 5 - 6 1503 67
GenBank mRNA AF145673.1 - 9 - 10 1532 67
BT099630.1 - 5 - 6 1173 67
BT133189.1 - 5 - 6 1096 67

View Full Product Details