Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- gem
- Entrez Gene ID
-
36064
- Gene Name
- gemini
- Gene Aliases
-
BcDNA:HL07889, CG11867, CG30011, CG3459, Dmel\CG30011, Dmel_CG30011, anon-AE003830.1, dCP2
- Chromosome Location
-
Chr.2R: 10124293 - 10137472 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050011
- Gene Symbol
- CG46319
- Entrez Gene ID
-
30522408
- Gene Name
- CG46319-PA
- Gene Aliases
-
Dmel\CG46319, Dmel_CG46319, gem-5'utr
- Chromosome Location
-
Chr.2R: 10124293 - 10137472 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0284235
View Full Product Details