Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAP
Entrez Gene ID
36084
Gene Name
CAP-PA
Gene Aliases
CAP/Vinexin, CG18408, CG18409, CG3451, DCAP, Dmel\CG18408, Dmel_CG18408, Rexin, cap, rexin
Chromosome Location
Chr.2R: 10269016 - 10307024 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033504
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144164.2 NP_001137636.1 2 - 3 577 67
NM_001382041.1 NP_001369073.1 2 - 3 786 67
NM_165757.3 NP_724910.1 3 - 4 598 67
NM_165758.2 NP_724911.1 2 - 3 786 67
GenBank mRNA BT003217.1 - 3 - 4 599 67
BT003435.1 - 2 - 3 558 67

View Full Product Details