Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oatp33Ea
Entrez Gene ID
34660
Gene Name
Organic anion transporting polypeptide 33Ea
Gene Aliases
CG5427, Dmel\CG5427, Dmel_CG5427, oatp 33Ea
Chromosome Location
Chr.2L: 12441401 - 12445924 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032433
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273488.1 NP_001260417.1 6 - 7 2481 87
NM_135724.4 NP_609568.3 6 - 7 2469 87
GenBank mRNA BT125052.1 - 6 - 7 2418 87

View Full Product Details