Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CAP
- Entrez Gene ID
-
36084
- Gene Name
- CAP-PA
- Gene Aliases
-
CAP/Vinexin, CG18408, CG18409, CG3451, DCAP, Dmel\CG18408, Dmel_CG18408, Rexin, cap, rexin
- Chromosome Location
-
Chr.2R: 10269016 - 10307024 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033504
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165761.2 |
NP_724914.1 |
1 - 2 |
640 |
78 |
View Full Product Details