Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbl6
Entrez Gene ID
36201
Gene Name
F-box and leucine-rich repeat protein 6
Gene Aliases
CG13213, Dmel\CG13213, Dmel_CG13213, FBXL6, fbl6
Chromosome Location
Chr.2R: 11285148 - 11289772 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033609
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136821.3 NP_610665.2 4 - 5 1998 103
NM_165815.2 NP_725019.1 4 - 5 1673 103
NM_165816.2 NP_725020.1 4 - 5 1396 103
GenBank mRNA BT023808.1 - 4 - 5 1649 103

View Full Product Details