Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or9a
- Entrez Gene ID
-
31975
- Gene Name
- Odorant receptor 9a
- Gene Aliases
-
9E.1, 9a, CG15302, DOR95, DmOr9a, Dmel\CG15302, Dmel_CG15302, OR9a, or9a
- Chromosome Location
-
Chr.X: 10458186 - 10459676 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030204
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078552.3 |
NP_511107.1 |
3 - 4 |
1121 |
101 |
View Full Product Details