Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- dsh
- Entrez Gene ID
-
32078
- Gene Name
- dishevelled
- Gene Aliases
-
CG1720, CG18361, DSH, DVL1-3, Dm Dsh, Dmel\CG18361, Dmel_CG18361, Dsh, Dvl, l(1)10Bd, l(1)G0267, l(1)G17, l(1)dsh
- Chromosome Location
-
Chr.X: 11355881 - 11358724 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000499
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078563.3 |
NP_511118.2 |
1 - 1 |
538 |
71 |
GenBank mRNA |
BT023781.1 |
- |
1 - 1 |
516 |
71 |
View Full Product Details