Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- hop
- Entrez Gene ID
-
32080
- Gene Name
- hopscotch
- Gene Aliases
-
4, CG1594, Dm JAK, DmHD-160, Dmel\CG1594, Dmel_CG1594, HD-160, HOP, Hop, Hop1, Hop2, JAK, Jak, L4, Tum, Tum-1, d-jak, hop/JAK, jak, l(1)10Be, l(1)G18, l(1)L4, l(1)hop, msvl
- Chromosome Location
-
Chr.X: 11360930 - 11368098 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004864
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078564.3 |
NP_511119.2 |
9 - 10 |
3974 |
69 |
GenBank mRNA |
BT006331.1 |
- |
9 - 10 |
3974 |
69 |
View Full Product Details