Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PhKgamma
Entrez Gene ID
32120
Gene Name
Phosphorylase kinase gamma
Gene Aliases
CG1830, DPhK-gamma, Dmel\CG1830, Dmel_CG1830, EP779, ESTS:194B3T, PhKg, Phkgamma, anon-sts6, phkgamma CES
Chromosome Location
Chr.X: 11695137 - 11704394 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011754
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298204.1 NP_001285133.1 7 - 8 1091 93
NM_001298205.1 NP_001285134.1 7 - 8 1297 93
NM_078574.4 NP_511129.3 7 - 8 1075 93
NM_167296.3 NP_727548.1 7 - 8 1196 93
NM_167297.3 NP_727549.2 7 - 8 1463 93
GenBank mRNA AY051575.1 - 7 - 8 1078 93
AY051635.1 - 7 - 8 1202 93

View Full Product Details