Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CkIalpha
Entrez Gene ID
32221
Gene Name
Casein kinase Ialpha
Gene Aliases
CG2028, CK I, CK1, CK1a, CK1alpha, CKI, CKI alpha, CKIa, CKIalpha, Ck1alpha, CkI alpha, CkIa, Dmel\CG2028, Dmel_CG2028, PKA-C, anon-WO03040301.93, anon-WO03040301.95, ck1a, ck1alpha, ckI, ckIalpha, dmCK1, dmckI, l(1)G0492
Chromosome Location
Chr.X: 12653626 - 12657834 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015024
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169266.3 NP_001162737.1 3 - 4 1285 90
NM_001298234.1 NP_001285163.1 3 - 4 1459 90
NM_001298235.1 NP_001285164.1 3 - 4 1050 90
NM_078585.6 NP_511140.1 3 - 4 1285 90
NM_167331.4 NP_727631.1 3 - 4 1186 90
NM_167332.3 NP_727632.1 3 - 4 1050 90
GenBank mRNA AY069346.1 - 3 - 4 1292 90
U55848.2 - 3 - 4 1229 90

View Full Product Details