Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rdgB
Entrez Gene ID
32340
Gene Name
retinal degeneration B
Gene Aliases
CG11111, Dmel\CG11111, Dmel_CG11111, DrdgB, DrdgBalpha, RDGBalpha, RdgB, RdgBalpha, hypoF, ota1, rdbB(ota1), rdgBalpha, x36
Chromosome Location
Chr.X: 13762752 - 13779488 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003218
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014740.2 NP_001014740.1 2 - 3 449 99
NM_001014741.3 NP_001014741.1 3 - 4 507 99
NM_001169277.2 NP_001162748.1 2 - 3 449 99
NM_001169278.3 NP_001162749.1 2 - 3 449 99
NM_001169279.3 NP_001162750.1 2 - 3 449 99
NM_001169280.3 NP_001162751.1 2 - 3 449 99
NM_001272605.2 NP_001259534.1 3 - 4 530 99
NM_001272606.1 NP_001259535.1 2 - 3 564 99
NM_001298300.1 NP_001285229.1 2 - 3 449 99
NM_078594.3 NP_511149.2 1 - 2 493 99
NM_167382.3 NP_727733.1 2 - 3 487 99
GenBank mRNA AY051422.1 - 2 - 3 469 99

View Full Product Details