Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rdgB
Entrez Gene ID
32340
Gene Name
retinal degeneration B
Gene Aliases
CG11111, Dmel\CG11111, Dmel_CG11111, DrdgB, DrdgBalpha, RDGBalpha, RdgB, RdgBalpha, hypoF, ota1, rdbB(ota1), rdgBalpha, x36
Chromosome Location
Chr.X: 13762752 - 13779488 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003218
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014740.2 NP_001014740.1 9 - 10 2958 75
NM_001014741.3 NP_001014741.1 10 - 11 3016 75
NM_001169277.2 NP_001162748.1 9 - 10 2958 75
NM_001169278.3 NP_001162749.1 9 - 10 2958 75
NM_001169280.3 NP_001162751.1 10 - 11 2997 75
NM_001272605.2 NP_001259534.1 11 - 12 3078 75
NM_001272606.1 NP_001259535.1 10 - 11 3112 75
NM_001298300.1 NP_001285229.1 9 - 10 2958 75
NM_078594.3 NP_511149.2 8 - 9 3002 75
NM_167382.3 NP_727733.1 9 - 10 2996 75
GenBank mRNA AY051422.1 - 9 - 10 2978 75

View Full Product Details