Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kat80
Entrez Gene ID
32581
Gene Name
katanin 80
Gene Aliases
CG13956, CG9910, Dmel\CG13956, Dmel_CG13956, Kat80, Katanin80
Chromosome Location
Chr.X: 16267007 - 16276180 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040207
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298388.1 NP_001285317.1 9 - 10 2572 108
NM_078639.3 NP_523363.2 10 - 11 2476 108
NM_167488.2 NP_727940.1 9 - 10 2575 108
GenBank mRNA BT044203.1 - 9 - 10 2556 108

View Full Product Details