Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
para
Entrez Gene ID
32619
Gene Name
paralytic
Gene Aliases
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0285944
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042815.2 NP_001036280.1 17 - 18 2828 79
NM_001042816.2 NP_001036281.1 17 - 18 2828 79
NM_001144733.2 NP_001138205.1 16 - 17 2771 79
NM_001201671.1 NP_001188600.1 17 - 18 2828 79
NM_001201672.1 NP_001188601.1 17 - 18 2828 79
NM_001201673.1 NP_001188602.1 16 - 17 2765 79
NM_001201674.1 NP_001188603.1 16 - 17 2795 79
NM_001201675.1 NP_001188604.1 16 - 17 2741 79
NM_001201676.1 NP_001188605.1 17 - 18 2804 79
NM_001201677.1 NP_001188606.1 17 - 18 2732 79
NM_001201678.1 NP_001188607.1 16 - 17 2765 79
NM_001201679.1 NP_001188608.1 17 - 18 2804 79
NM_001201680.1 NP_001188609.1 16 - 17 2765 79
NM_001201681.1 NP_001188610.1 16 - 17 2741 79
NM_001201682.1 NP_001188611.1 17 - 18 2828 79
NM_001201683.1 NP_001188612.1 17 - 18 2732 79
NM_001201684.1 NP_001188613.1 16 - 17 2741 79
NM_001201685.1 NP_001188614.1 16 - 17 2765 79
NM_001201686.1 NP_001188615.1 17 - 18 2804 79
NM_001201687.1 NP_001188616.1 17 - 18 2828 79
NM_001201688.1 NP_001188617.1 16 - 17 2765 79
NM_001201689.1 NP_001188618.1 16 - 17 2741 79
NM_001201690.1 NP_001188619.1 17 - 18 2804 79
NM_001201691.1 NP_001188620.1 17 - 18 2804 79
NM_001201692.1 NP_001188621.1 17 - 18 2732 79
NM_001201693.1 NP_001188622.1 17 - 18 2756 79
NM_001201694.1 NP_001188623.1 17 - 18 2732 79
NM_001201695.2 NP_001188624.1 17 - 18 2756 79
NM_001201696.1 NP_001188625.1 15 - 16 2732 79
NM_001201697.1 NP_001188626.1 17 - 18 2828 79
NM_001201698.1 NP_001188627.1 16 - 17 2795 79
NM_001201699.1 NP_001188628.1 15 - 16 2732 79
NM_001201700.1 NP_001188629.1 16 - 17 2795 79
NM_001201701.1 NP_001188630.1 15 - 16 2732 79
NM_001201702.1 NP_001188631.1 16 - 17 2795 79
NM_001201703.3 NP_001188632.1 17 - 18 3318 79
NM_001201704.1 NP_001188633.1 15 - 16 2732 79
NM_001201705.1 NP_001188634.1 16 - 17 2723 79
NM_001201706.1 NP_001188635.1 17 - 18 2828 79
NM_001201707.1 NP_001188636.1 17 - 18 2804 79
NM_001201708.1 NP_001188637.1 16 - 17 2765 79
NM_001201709.1 NP_001188638.1 16 - 17 2741 79
NM_001201710.1 NP_001188639.1 13 - 14 2511 79
NM_001201711.1 NP_001188640.1 15 - 16 2666 79
NM_001201712.1 NP_001188641.1 14 - 15 2603 79
NM_001201713.1 NP_001188642.1 14 - 15 2603 79
NM_001201714.1 NP_001188643.1 14 - 15 2579 79
NM_001201715.1 NP_001188644.1 15 - 16 2660 79
NM_001201716.1 NP_001188645.1 16 - 17 2771 79
NM_001201717.1 NP_001188646.1 16 - 17 2795 79
NM_001201718.1 NP_001188647.1 15 - 16 2732 79
NM_001201719.1 NP_001188648.1 16 - 17 2795 79
NM_001201720.1 NP_001188649.1 16 - 17 2795 79
NM_001201721.1 NP_001188650.1 15 - 16 2636 79
NM_001201722.1 NP_001188651.1 16 - 17 2795 79
NM_001201723.1 NP_001188652.1 16 - 17 2771 79
NM_001272690.1 NP_001259619.1 17 - 18 2828 79
NM_001298403.1 NP_001285332.1 17 - 18 2981 79
NM_001298404.1 NP_001285333.1 16 - 17 2792 79
NM_078647.4 NP_523371.2 17 - 18 2828 79

View Full Product Details