Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
para
Entrez Gene ID
32619
Gene Name
paralytic
Gene Aliases
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0285944
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042815.2 NP_001036280.1 30 - 31 5658 98
NM_001042816.2 NP_001036281.1 30 - 31 5658 98
NM_001144733.2 NP_001138205.1 29 - 30 5607 98
NM_001201671.1 NP_001188600.1 30 - 31 5589 98
NM_001201672.1 NP_001188601.1 30 - 31 5589 98
NM_001201673.1 NP_001188602.1 29 - 30 5526 98
NM_001201674.1 NP_001188603.1 29 - 30 5556 98
NM_001201675.1 NP_001188604.1 29 - 30 5502 98
NM_001201676.1 NP_001188605.1 30 - 31 5565 98
NM_001201677.1 NP_001188606.1 30 - 31 5493 98
NM_001201678.1 NP_001188607.1 29 - 30 5526 98
NM_001201679.1 NP_001188608.1 30 - 31 5565 98
NM_001201680.1 NP_001188609.1 29 - 30 5565 98
NM_001201681.1 NP_001188610.1 29 - 30 5541 98
NM_001201682.1 NP_001188611.1 30 - 31 5628 98
NM_001201683.1 NP_001188612.1 30 - 31 5493 98
NM_001201684.1 NP_001188613.1 29 - 30 5502 98
NM_001201685.1 NP_001188614.1 29 - 30 5565 98
NM_001201686.1 NP_001188615.1 30 - 31 5595 98
NM_001201687.1 NP_001188616.1 30 - 31 5619 98
NM_001201688.1 NP_001188617.1 29 - 30 5556 98
NM_001201689.1 NP_001188618.1 29 - 30 5571 98
NM_001201690.1 NP_001188619.1 30 - 31 5640 98
NM_001201691.1 NP_001188620.1 30 - 31 5679 98
NM_001201692.1 NP_001188621.1 30 - 31 5532 98
NM_001201693.1 NP_001188622.1 30 - 31 5517 98
NM_001201694.1 NP_001188623.1 30 - 31 5523 98
NM_001201695.2 NP_001188624.1 30 - 31 5517 98
NM_001201696.1 NP_001188625.1 28 - 29 5523 98
NM_001201697.1 NP_001188626.1 30 - 31 5589 98
NM_001201698.1 NP_001188627.1 29 - 30 5595 98
NM_001201699.1 NP_001188628.1 28 - 29 5532 98
NM_001201700.1 NP_001188629.1 29 - 30 5586 98
NM_001201701.1 NP_001188630.1 28 - 29 5523 98
NM_001201702.1 NP_001188631.1 29 - 30 5661 98
NM_001201703.3 NP_001188632.1 30 - 31 6079 98
NM_001201704.1 NP_001188633.1 28 - 29 5598 98
NM_001201705.1 NP_001188634.1 29 - 30 5514 98
NM_001201706.1 NP_001188635.1 30 - 31 5694 98
NM_001201707.1 NP_001188636.1 30 - 31 5604 98
NM_001201708.1 NP_001188637.1 29 - 30 5556 98
NM_001201709.1 NP_001188638.1 29 - 30 5532 98
NM_001201710.1 NP_001188639.1 26 - 27 5416 98
NM_001201711.1 NP_001188640.1 28 - 29 5466 98
NM_001201712.1 NP_001188641.1 27 - 28 5433 98
NM_001201713.1 NP_001188642.1 25 - 26 4774 98
NM_001201714.1 NP_001188643.1 25 - 26 4750 98
NM_001201715.1 NP_001188644.1 28 - 29 5451 98
NM_001201716.1 NP_001188645.1 29 - 30 5562 98
NM_001201717.1 NP_001188646.1 29 - 30 5661 98
NM_001201718.1 NP_001188647.1 28 - 29 5598 98
NM_001201719.1 NP_001188648.1 29 - 30 5661 98
NM_001201720.1 NP_001188649.1 28 - 29 5463 98
NM_001201721.1 NP_001188650.1 24 - 25 4521 98
NM_001201722.1 NP_001188651.1 28 - 29 5433 98
NM_001201723.1 NP_001188652.1 28 - 29 5474 98
NM_001272690.1 NP_001259619.1 30 - 31 5658 98
NM_001298403.1 NP_001285332.1 30 - 31 5811 98
NM_001298404.1 NP_001285333.1 28 - 29 5430 98
NM_078647.4 NP_523371.2 30 - 31 5658 98

View Full Product Details