Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
if
Entrez Gene ID
32661
Gene Name
inflated
Gene Aliases
CG9623, CT27194, Dmel\CG9623, Dmel_CG9623, If, ItgaPS2, PS 2, PS2, PS2(c), PS2(m8), PS2(m8)alpha, PS2alpha, PSalpha2, aPS2, alpha-PS2, alpha2Int, alphaPS2, alphaPS2 integrin, alphaPS2C, alphaPS2m8, alpha[[PS2(ms8)]], alpha[[PS2]], in, inf
Chromosome Location
Chr.X: 16752189 - 16783434 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001250
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169306.2 NP_001162777.1 9 - 10 2246 59
NM_001169307.1 NP_001162778.1 9 - 10 2381 59
NM_078654.2 NP_523378.2 9 - 10 2287 59
NM_167544.2 NP_728021.2 8 - 9 2212 59

View Full Product Details