Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
baz
Entrez Gene ID
32703
Gene Name
bazooka
Gene Aliases
BAZ, Baz, Baz/Par-3, Baz/Par3, Bazooka, CG5055, D-Par3, D-par3, Dmel\CG5055, Dmel_CG5055, PAR-3, PAR3, Par-3, Par3, dPar-3, l(1)G0484, par-3, par3
Chromosome Location
Chr.X: 17160006 - 17200728 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000163
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042818.4 NP_001036283.1 7 - 8 8414 81
NM_001298435.2 NP_001285364.1 5 - 6 4460 81
NM_001298436.2 NP_001285365.1 7 - 8 8414 81
NM_001347807.1 NP_001334669.1 5 - 6 4460 81
GenBank mRNA BT032834.1 - 5 - 6 3531 81

View Full Product Details