Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Dhc16F
- Entrez Gene ID
-
32785
- Gene Name
- Dynein heavy chain at 16F
- Gene Aliases
-
CG7092, DHC, DHC9, Dhc, Dmel\CG7092, Dmel_CG7092, atoll, cvl-6, dhc
- Chromosome Location
-
Chr.X: 18064483 - 18078472 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0283476
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078670.3 |
NP_523394.1 |
2 - 3 |
1158 |
64 |
View Full Product Details