Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dhc16F
Entrez Gene ID
32785
Gene Name
Dynein heavy chain at 16F
Gene Aliases
CG7092, DHC, DHC9, Dhc, Dmel\CG7092, Dmel_CG7092, atoll, cvl-6, dhc
Chromosome Location
Chr.X: 18064483 - 18078472 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283476
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078670.3 NP_523394.1 9 - 10 3255 57

View Full Product Details