Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bx
Entrez Gene ID
32846
Gene Name
Beadex
Gene Aliases
Bd, CG10548, CG15048, CG44425, CG6500, DLMO, Dlmo, Dmel\CG44425, Dmel_CG44425, LMO, Lmo, Ptd, Rho, beadex/dLMO, bx, dLMO, dLmo, dlmo, dttg, fliH, hdp, hdp-a, hld
Chromosome Location
Chr.X: 18515428 - 18572809 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265598
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272758.1 NP_001259687.1 2 - 3 1204 66
NM_001272759.2 NP_001259688.2 3 - 4 1019 66
NM_078678.4 NP_523402.4 2 - 3 761 66
NM_167625.3 NP_728184.1 2 - 3 1204 66
NM_176758.3 NP_788931.1 3 - 4 1040 66
GenBank mRNA AY060902.1 - 2 - 3 1136 66
BT004844.1 - 2 - 3 730 66

View Full Product Details