Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pvf1
Entrez Gene ID
32876
Gene Name
PDGF- and VEGF-related factor 1
Gene Aliases
CG7103, DmVEGF-1, Dmel\CG7103, Dmel_CG7103, EP1624, PVF, PVF1, PVFs, Pvf, Pvf-1, VEGF, VEGF2-B, Vegf17E, Vegf17e-a, Vegf17e-b, l(1)G0146, pvf1, vegf1
Chromosome Location
Chr.X: 18832682 - 18842901 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030964
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169321.2 NP_001162792.1 5 - 6 1119 63
NM_078683.3 NP_523407.1 5 - 6 1211 63
GenBank mRNA AJ401391.1 - 5 - 6 1097 63
AY051751.1 - 5 - 6 1189 63
AY079186.1 - 5 - 6 1098 63
AY079188.1 - 5 - 6 1131 63

View Full Product Details