Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Obp19d
Entrez Gene ID
33040
Gene Name
Odorant-binding protein 19d
Gene Aliases
19d, BcDNA:HL07789, BcDNA:RH68082, CG1668, DmelObp19d, Dmel\CG1668, Dmel_CG1668, PB-PRP2, PBP2, PBPRP-2, PBPRP2, Pbprp2, pbprp-2, pbprp2
Chromosome Location
Chr.X: 20419613 - 20420979 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011280
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078697.3 NP_523421.2 3 - 4 906 70
NM_176767.2 NP_788940.1 3 - 4 429 70
GenBank mRNA AY118484.1 - 4 - 5 925 70
KX532139.1 - 3 - 4 381 70

View Full Product Details