Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GABA-B-R3
Entrez Gene ID
33248
Gene Name
metabotropic GABA-B receptor subtype 3
Gene Aliases
CG3022, D-GABA[[B]]R3, D-Gaba3, Dmel\CG3022, Dmel_CG3022, GABA B R3, GABA-BR3, GABA[[B]]-R3, GABA[[B]]R3, GABAb R3, GB3, anon-WO0170980.13, anon-WO0170980.14
Chromosome Location
Chr.2L: 749945 - 762399 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031275
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258897.1 NP_001245826.1 7 - 8 2529 59
NM_001298623.1 NP_001285552.1 6 - 7 2599 59
NM_001298624.1 NP_001285553.1 6 - 7 2432 59
NM_001298625.1 NP_001285554.1 7 - 8 2529 59
NM_078723.3 NP_523447.2 6 - 7 2265 59
NM_164387.2 NP_722648.2 7 - 8 2790 59

View Full Product Details