Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ninaC
- Entrez Gene ID
-
34012
- Gene Name
- neither inactivation nor afterpotential C
- Gene Aliases
-
2.2, CG 5125, CG5125, CG54125, CT16120, CT42491, DRONINAC, Dm NinaC, Dmel\CG5125, Dmel_CG5125, Myo1B, NINA C, NINAC, Nina C, NinaC, ninac
- Chromosome Location
-
Chr.2L: 7377702 - 7384344 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002938
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078779.3 |
NP_523503.2 |
10 - 11 |
3430 |
84 |
View Full Product Details