Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tep2
Entrez Gene ID
34044
Gene Name
Thioester-containing protein 2
Gene Aliases
145970_at, 145971_at, CG18589, CG7052, Dmel\CG7052, Dmel_CG7052, T13, TEP II, TEP2, TEP2mel, Tep 2, Tep II, TepII, dTEPII, tepII
Chromosome Location
Chr.2L: 7693727 - 7701601 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041182
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078782.3 NP_523506.1 5 - 6 2282 97
GenBank mRNA AJ269539.1 - 5 - 6 2266 97
AY122084.1 - 5 - 6 2266 97

View Full Product Details