Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ttc19
- Entrez Gene ID
-
35172
- Gene Name
- Tetratricopeptide repeat domain 19
- Gene Aliases
-
CG15173, Dmel\CG15173, Dmel_CG15173, TTC19, cg15173
- Chromosome Location
-
Chr.2L: 19043535 - 19045148 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032744
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136090.3 |
NP_609934.3 |
3 - 4 |
1091 |
98 |
GenBank mRNA |
AY071630.1 |
- |
3 - 4 |
916 |
98 |
View Full Product Details