Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPA2
Entrez Gene ID
35364
Gene Name
Replication protein A2
Gene Aliases
153194_at, CG9273, Dmel\CG9273, Dmel_CG9273, RpA2, dRPA2
Chromosome Location
Chr.2L: 20924975 - 20926147 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032906
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273723.1 NP_001260652.1 3 - 4 763 74
NM_136233.3 NP_610077.1 3 - 4 763 74
GenBank mRNA AY069259.1 - 3 - 4 738 74

View Full Product Details