Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk13
Entrez Gene ID
3346226
Gene Name
pickpocket 13
Gene Aliases
CG14398, CG33508, Dmel\CG33508, Dmel_CG33508, PPK13
Chromosome Location
Chr.2L: 21086790 - 21088751 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053508
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014495.2 NP_001014495.1 2 - 3 309 79
GenBank mRNA AY226544.1 - 2 - 3 288 79

View Full Product Details