Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fmo-2
- Entrez Gene ID
-
35561
- Gene Name
- Flavin-containing monooxygenase 2
- Gene Aliases
-
CG3174, DmFMO-2, Dmel\CG3174, Dmel_CG3174, Dmfmo2, fmo2
- Chromosome Location
-
Chr.2R: 6642418 - 6645098 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033079
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136373.4 |
NP_610217.1 |
1 - 2 |
135 |
57 |
GenBank mRNA |
AY058349.1 |
- |
1 - 2 |
117 |
57 |
View Full Product Details