Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pgant3
- Entrez Gene ID
-
35627
- Gene Name
- Polypeptide N-Acetylgalactosaminyltransferase 3
- Gene Aliases
-
BcDNA:GH09147, CG4445, Dmel\CG4445, Dmel_CG4445, anon-WO0118547.119, pgant3
- Chromosome Location
-
Chr.2R: 7057654 - 7061732 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027558
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136412.4 |
NP_610256.1 |
4 - 5 |
954 |
79 |
GenBank mRNA |
AF145655.1 |
- |
4 - 5 |
928 |
79 |
View Full Product Details