Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Glo1
- Entrez Gene ID
-
35656
- Gene Name
- Glyoxalase 1
- Gene Aliases
-
CG1707, Dmel\CG1707, Dmel_CG1707, glo1
- Chromosome Location
-
Chr.2R: 7393546 - 7395932 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0283450
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136426.3 |
NP_610270.1 |
2 - 3 |
275 |
71 |
View Full Product Details