Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps13
Entrez Gene ID
35693
Gene Name
Vacuolar protein sorting 13
Gene Aliases
BEST:LD02743, CG2093, Dmel\CG2093, Dmel_CG2093
Chromosome Location
Chr.2R: 7566809 - 7580675 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033194
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273852.1 NP_001260781.1 27 - 28 9603 90
NM_136455.3 NP_610299.2 27 - 28 9487 90

View Full Product Details