Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dgp-1
Entrez Gene ID
37080
Gene Name
CG5729-PA
Gene Aliases
CG5729, Dmel\CG5729, Dmel_CG5729
Chromosome Location
Chr.2R: 18168413 - 18172607 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027836
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137458.3 NP_611302.1 3 - 4 613 71
NM_166274.2 NP_725760.1 3 - 4 705 71
GenBank mRNA BT133046.1 - 3 - 4 681 71

View Full Product Details