Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cora
Entrez Gene ID
37205
Gene Name
coracle
Gene Aliases
CG11949, COR, Cor, Cora, Coracle, D4.1, Dmel\CG11949, Dmel_CG11949, cor, l(2)k08713, mdcds_14526
Chromosome Location
Chr.2R: 19229063 - 19243751 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010434
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299678.1 NP_001286607.1 13 - 14 4999 66
NM_001299679.1 NP_001286608.1 14 - 15 5287 66
NM_079067.4 NP_523791.2 14 - 15 5293 66
NM_166336.2 NP_725864.1 13 - 14 2308 66
NM_166337.2 NP_725865.1 14 - 15 2866 66
NM_166338.3 NP_725866.1 13 - 14 2296 66
GenBank mRNA BT024976.1 - 13 - 14 2279 66
BT133208.1 - 13 - 14 2294 66

View Full Product Details