Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EndoB
Entrez Gene ID
37218
Gene Name
Endophilin B
Gene Aliases
CG9834, D-EndoB, D-endoB, Dmel\CG9834, Dmel_CG9834, endoB
Chromosome Location
Chr.2R: 19307447 - 19310665 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034433
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137566.4 NP_611410.1 5 - 6 1161 94
NM_166340.3 NP_725873.1 4 - 5 1125 94
GenBank mRNA AJ437142.1 - 4 - 5 1081 94
AY071608.1 - 5 - 6 1188 94
BT044418.1 - 4 - 5 1110 94

View Full Product Details