Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hil
Entrez Gene ID
37328
Gene Name
Hillarin
Gene Aliases
CG10439, CG13435, CG30147, D-hil, Dmel\CG30147, Dmel_CG30147
Chromosome Location
Chr.2R: 20606881 - 20618217 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050147
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137649.3 NP_611493.2 8 - 9 2614 74
NM_166402.2 NP_725980.1 8 - 9 2677 74

View Full Product Details