Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jbug
Entrez Gene ID
43997
Gene Name
jitterbug
Gene Aliases
AAF46896, CG11605, CG13525, CG30092, CG3550, Dmel\CG30092, Dmel_CG30092, Jbug
Chromosome Location
Chr.2R: 22717321 - 22740459 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028371
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169787.2 NP_001163258.1 4 - 5 1586 52
NM_001274210.1 NP_001261139.1 11 - 12 4640 52
NM_001274211.1 NP_001261140.1 12 - 13 4988 52
NM_166546.3 NP_726234.3 11 - 12 5027 52

View Full Product Details