Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jbug
Entrez Gene ID
43997
Gene Name
jitterbug
Gene Aliases
AAF46896, CG11605, CG13525, CG30092, CG3550, Dmel\CG30092, Dmel_CG30092, Jbug
Chromosome Location
Chr.2R: 22717321 - 22740459 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028371
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169787.2 NP_001163258.1 7 - 8 5323 80
NM_001274210.1 NP_001261139.1 14 - 15 8377 80
NM_001274211.1 NP_001261140.1 15 - 16 8725 80
NM_166546.3 NP_726234.3 14 - 15 8764 80

View Full Product Details